tert-Butyl (2-fluoroethyl)carbamate 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC48733-1G
PC48733-250MG
APOSPC48733-1GEA
18620
CZK
APOSPC48733-1G
APOSPC48733-250MG
tert-Butyl (2-fluoroethyl)carbamate 95%
tert-Butyl (2-fluoroethyl)carbamate
Vzorec:
C₇H₁₄FNO₂ MW: 163,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22374847 CAS číslo: 178181-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...