Tisk…

2-Fluoro-4-iodo-5-(methoxymethoxy)pyridine

Dodavatel: Apollo Scientific
Varování

PC48727-1G PC48727-500MG PC48727-250MG
APOSPC48727-1GEA 2240 CZK
APOSPC48727-1G APOSPC48727-500MG APOSPC48727-250MG
2-Fluoro-4-iodo-5-(methoxymethoxy)pyridine
2-Fluoro-4-iodo-5-(methoxymethoxy)pyridine
Vzorec: C₇H₇FINO₂
MW: 283,039 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22123918
CAS číslo: 1034467-27-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR