Tisk…

1-(4-Bromo-2-fluorophenyl)-3-(2-chloroethyl)urea

Dodavatel: Apollo Scientific
Varování

PC48675-1G PC48675-250MG
APOSPC48675-1GEA 5920 CZK
APOSPC48675-1G APOSPC48675-250MG
1-(4-Bromo-2-fluorophenyl)-3-(2-chloroethyl)urea
1-(4-Bromo-2-fluorophenyl)-3-(2-chloroethyl)urea
Vzorec: C₉H₉BrClFN₂O
MW: 295,536 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31372485
CAS číslo: 160132-69-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR