Tisk…

1-(3-Bromo-4-fluorophenyl)-3-(2-chloroethyl)urea

Dodavatel: Apollo Scientific
Varování

PC48674-1G PC48674-250MG
APOSPC48674-1GEA 5920 CZK
APOSPC48674-1G APOSPC48674-250MG
1-(3-Bromo-4-fluorophenyl)-3-(2-chloroethyl)urea
1-(3-Bromo-4-fluorophenyl)-3-(2-chloroethyl)urea
Vzorec: C₉H₉BrClFN₂O
Storage Temperature: Ambient
MDL Number: MFCD31372792
CAS číslo: 2168111-87-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR