1-(4-Bromo-3-fluorophenyl)-3-(2-chloroethyl)urea
Dodavatel: Apollo Scientific
|
Varování
|
PC48673-1G
PC48673-250MG
APOSPC48673-1GEA
5920
CZK
APOSPC48673-1G
APOSPC48673-250MG
1-(4-Bromo-3-fluorophenyl)-3-(2-chloroethyl)urea
1-(4-Bromo-3-fluorophenyl)-3-(2-chloroethyl)urea
Vzorec:
C₉H₉BrClFN₂O MW: 295,536 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31372515 CAS číslo: 2089312-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...