Tisk…

Methyl 4-bromo-3-chloro-2,6-difluorobenzoate 95%

Dodavatel: Apollo Scientific
Varování

PC48663-1G PC48663-250MG PC48663-5G
APOSPC48663-1GEA 1660 CZK
APOSPC48663-1G APOSPC48663-250MG APOSPC48663-5G
Methyl 4-bromo-3-chloro-2,6-difluorobenzoate 95%
Methyl 4-bromo-3-chloro-2,6-difluorobenzoate
Vzorec: C₈H₄BrClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD24108605
CAS číslo: 1500268-85-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR