Tisk…

2,2,2-Trifluoro-1-[3-nitro-5-(trifluoromethyl)phenyl]ethanone 95%

Dodavatel: Apollo Scientific
Varování

PC48662-1G PC48662-250MG PC48662-5G
APOSPC48662-1GEA 992 CZK
APOSPC48662-1G APOSPC48662-250MG APOSPC48662-5G
2,2,2-Trifluoro-1-[3-nitro-5-(trifluoromethyl)phenyl]ethanone 95%
2,2,2-Trifluoro-1-[3-nitro-5-(trifluoromethyl)phenyl]ethanone
Vzorec: C₉H₃F₆NO₃
MW: 287,115 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30295050
CAS číslo: 76911-72-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR