Tisk…

2-(3,4-Diamino-5-fluorophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 95%

Dodavatel: Apollo Scientific
Varování

PC48660-1G PC48660-250MG PC48660-5G
APOSPC48660-1GEA 1660 CZK
APOSPC48660-1G APOSPC48660-250MG APOSPC48660-5G
2-(3,4-Diamino-5-fluorophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 95%
2-(3,4-Diamino-5-fluorophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Vzorec: C₉H₇F₇N₂O
MW: 292.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32066760
CAS číslo: 2366994-07-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR