Tisk…

Methyl 4-(chlorosulphonyl)-3-fluorobenzoate

Dodavatel: Apollo Scientific
Nebezpečí

PC48658-1G PC48658-250MG PC48658-5G
APOSPC48658-1GEA 14880 CZK
APOSPC48658-1G APOSPC48658-250MG APOSPC48658-5G
Methyl 4-(chlorosulphonyl)-3-fluorobenzoate
Methyl 4-(chlorosulphonyl)-3-fluorobenzoate
Vzorec: C₈H₆ClFO₄S
MW: 252,647 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628063
CAS číslo: 1219687-20-3
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR