3-(Trifluoromethyl)pyridine-2-carboxamidine hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
PC48655-1G
PC48655-250MG
APOSPC48655-1GEA
10550
CZK
APOSPC48655-1G
APOSPC48655-250MG
3-(Trifluoromethyl)pyridine-2-carboxamidine hydrochloride
3-(Trifluoromethyl)pyridine-2-carboxamidine hydrochloride
Vzorec:
C₇H₆F₃N₃·ClH MW: 225.599 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09028472 CAS číslo: 1179361-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...