2-Hydroxy-6-(thien-2-yl)-4-(trifluoromethyl)pyridine-3-carbonitrile
Dodavatel: Apollo Scientific
PC4865-10G
PC4865-1G
PC4865-5G
APOSPC4865-10GEA
10550
CZK
APOSPC4865-10G
APOSPC4865-1G
APOSPC4865-5G
2-Hydroxy-6-(thien-2-yl)-4-(trifluoromethyl)pyridine-3-carbonitrile
2-Hydroxy-6-(thien-2-yl)-4-(trifluoromethyl)pyridine-3-carbonitrile
Vzorec:
C₁₁H₅F₃N₂OS MW: 270,23 g/mol Bod tání: 278 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052654 CAS číslo: 3335-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...