Tisk…

6-Bromo-2-fluoro-3-methylbenzaldehyde 97%

Dodavatel: Apollo Scientific

PC48649-1G PC48649-5G
APOSPC48649-1GEA 3310 CZK
APOSPC48649-1G APOSPC48649-5G
6-Bromo-2-fluoro-3-methylbenzaldehyde 97%
6-Bromo-2-fluoro-3-methylbenzaldehyde
Vzorec: C₈H₆BrFO
Storage Temperature: Refrigerator
MDL Number: MFCD11520668
CAS číslo: 1114809-22-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR