Tisk…

2,3-Dibromo-6-fluorobenzaldehyde 97%

Dodavatel: Apollo Scientific

PC48648-1G PC48648-5G
APOSPC48648-1GEA 5200 CZK
APOSPC48648-1G APOSPC48648-5G
2,3-Dibromo-6-fluorobenzaldehyde 97%
2,3-Dibromo-6-fluorobenzaldehyde
Vzorec: C₇H₃Br₂FO
MW: 281,905 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520662
CAS číslo: 1114809-15-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR