(R)-2-Chloro-1-(4-fluorophenyl)ethanol 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC48642-1G
PC48642-250MG
APOSPC48642-1GEA
1450
CZK
APOSPC48642-1G
APOSPC48642-250MG
(R)-2-Chloro-1-(4-fluorophenyl)ethanol 98%
(R)-2-Chloro-1-(4-fluorophenyl)ethanol
Vzorec:
C₈H₈ClFO MW: 174,6 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08064295 CAS číslo: 126534-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...