Tisk…

2,2'-(5-Bromo-2-fluorobenzene-1,3-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

Dodavatel: Apollo Scientific

PC48620-1G PC48620-5G
APOSPC48620-1GEA 8100 CZK
APOSPC48620-1G APOSPC48620-5G
2,2'-(5-Bromo-2-fluorobenzene-1,3-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)
2,2'-(5-Bromo-2-fluorobenzene-1,3-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)
Vzorec: C₁₈H₂₆B₂BrFO₄
Storage Temperature: Ambient
MDL Number: MFCD12026099
CAS číslo: 942069-49-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR