Tisk…

4-Bromo-2-chloro-6-(trifluoromethyl)pyridine

Dodavatel: Apollo Scientific

PC48601-1G PC48601-250MG
APOSPC48601-1GEA 25460 CZK
APOSPC48601-1G APOSPC48601-250MG
4-Bromo-2-chloro-6-(trifluoromethyl)pyridine
4-Bromo-2-chloro-6-(trifluoromethyl)pyridine
Vzorec: C₆H₂BrClF₃N
MW: 260,439 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189905
CAS číslo: 1196153-93-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR