Tisk…

5-Fluorobenzo[b]furan 95%

Dodavatel: Apollo Scientific

Synonyms: 5-Fluorobenzofuran, 5-Fluoro-1-benzofuran

PC48599-1G PC48599-250MG
APOSPC48599-1GEA 7250 CZK
APOSPC48599-1G APOSPC48599-250MG
5-Fluorobenzo[b]furan 95%
5-Fluorobenzo[b]furan
Vzorec: C₈H₅FO
MW: 136,123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08236757
CAS číslo: 24410-59-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR