Tisk…

4-Bromo-2-fluoro-3-methylbenzonitrile

Dodavatel: Apollo Scientific
Varování

PC48590-1G PC48590-250MG PC48590-5G
APOSPC48590-1GEA 11580 CZK
APOSPC48590-1G APOSPC48590-250MG APOSPC48590-5G
4-Bromo-2-fluoro-3-methylbenzonitrile
4-Bromo-2-fluoro-3-methylbenzonitrile
Vzorec: C₈H₅BrFN
MW: 214,034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23707391
CAS číslo: 1114546-30-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR