Tisk…

1H,1H,2H,2H-Perfluoroheptylamine

Dodavatel: Apollo Scientific
Nebezpečí

PC48573-1G PC48573-5G PC48573-250MG
APOSPC48573-1GEA 8600 CZK
APOSPC48573-1G APOSPC48573-5G APOSPC48573-250MG
1H,1H,2H,2H-Perfluoroheptylamine
1H,1H,2H,2H-Perfluoroheptylamine
Vzorec: C₇H₆F₁₁N
MW: 313,112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20489384
CAS číslo: 1346521-42-3
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR