Methyl 6-bromo-3-chloro-2-fluorobenzoate 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC48559-1G
PC48559-250MG
PC48559-5G
APOSPC48559-1GEA
9330
CZK
APOSPC48559-1G
APOSPC48559-250MG
APOSPC48559-5G
Methyl 6-bromo-3-chloro-2-fluorobenzoate 95%
Methyl 6-bromo-3-chloro-2-fluorobenzoate
Vzorec:
C₈H₅BrClFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD27978532 CAS číslo: 1505850-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...