Tisk…
(1R)-1-(4-Chloro-3-fluorophenyl)ethylamine hydrochloride 98%
Dodavatel: Apollo Scientific
Synonyms: (R)-1-(4-Chloro-3-fluorophenyl)ethanamine hydrochloride, (R)-4-Chloro-3-fluoro-α-methylbenzylamine hydrochloride
PC48555-100MG PC48555-250MG PC48555-1G
APOSPC48555-100MGEA 5590 CZK
APOSPC48555-100MG APOSPC48555-250MG APOSPC48555-1G
(1R)-1-(4-Chloro-3-fluorophenyl)ethylamine hydrochloride 98%
(1R)-1-(4-Chloro-3-fluorophenyl)ethylamine hydrochloride
Vzorec: C₈H₉ClFN·ClH
MW: 210,076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547495
CAS číslo: 1253790-80-5