Tisk…

2-(Difluoromethyl)benzonitrile

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Cyanobenzal fluoride, α,α-Difluoro-o-tolunitrile

PC48525-1G PC48525-5G
APOSPC48525-1GEA 1580 CZK
APOSPC48525-1G APOSPC48525-5G
2-(Difluoromethyl)benzonitrile
2-(Difluoromethyl)benzonitrile
Vzorec: C₈H₅F₂N
MW: 153,129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21606585
CAS číslo: 799814-30-5
UN: 3276
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR