2-(2,5-Dimethylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1,4-Dimethyl-2-[perfluoro(2-hydroxyprop-2-yl)]benzene, 2-(1,1,1,3,3,3-Hexafluoro-2-hydroxyprop-2-yl)-p-xylene
PC4852-1G
PC4852-250MG
APOSPC4852-1GEA
1660
CZK
APOSPC4852-1G
APOSPC4852-250MG
2-(2,5-Dimethylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 97%
2-(2,5-Dimethylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Vzorec:
C₁₁H₁₀F₆O MW: 272.187 g/mol Bod varu: 82…83 °C (10 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00041490 CAS číslo: 28180-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...