Tisk…

6-Bromo-3-chloro-2-fluorophenacyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

PC48480-1G PC48480-250MG
APOSPC48480-1GEA 10910 CZK
APOSPC48480-1G APOSPC48480-250MG
6-Bromo-3-chloro-2-fluorophenacyl bromide
6-Bromo-3-chloro-2-fluorophenacyl bromide
Vzorec: C₈H₄Br₂ClFO
Storage Temperature: Ambient
MDL Number: MFCD28096621
CAS číslo: 1804382-46-4
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR