Methyl 6-amino-2-fluoro-3-nitrobenzoate, tech
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Fluoro-2-(methoxycarbonyl)-4-nitroaniline
PC48472-1G
PC48472-250MG
APOSPC48472-1GEA
12200
CZK
APOSPC48472-1G
APOSPC48472-250MG
Methyl 6-amino-2-fluoro-3-nitrobenzoate, tech
Methyl 6-amino-2-fluoro-3-nitrobenzoate
Vzorec:
C₈H₇FN₂O₄ MW: 214,151 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28397885 CAS číslo: 1805398-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...