Tisk…

6-Bromo-3-chloro-2-fluorobenzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Chloro-α,6-dibromo-2-fluorotoluene, 1-Bromo-2-(bromomethyl)-4-chloro-3-fluorobenzene

PC48461-1G PC48461-250MG
APOSPC48461-1GEA 20500 CZK
APOSPC48461-1G APOSPC48461-250MG
6-Bromo-3-chloro-2-fluorobenzyl bromide
6-Bromo-3-chloro-2-fluorobenzyl bromide
Vzorec: C₇H₄Br₂ClF
MW: 302,366 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28096619
CAS číslo: 1820741-85-2
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR