6-Bromo-3-chloro-2-fluoroaniline 90%
Dodavatel: Apollo Scientific
|
Varování
|
PC48453-1G
PC48453-250MG
PC48453-100MG
APOSPC48453-1GEA
13000
CZK
APOSPC48453-1G
APOSPC48453-250MG
APOSPC48453-100MG
6-Bromo-3-chloro-2-fluoroaniline 90%
6-Bromo-3-chloro-2-fluoroaniline
Vzorec:
C₆H₄BrClFN MW: 224,458 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28096616 CAS číslo: 1515343-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...