Tisk…

Tris(2,2,3,3,3-pentafluoropropyl) borate

Dodavatel: Apollo Scientific
Varování

PC48448-5G PC48448-250MG PC48448-1G
APOSPC48448-5GEA 21660 CZK
APOSPC48448-5G APOSPC48448-250MG APOSPC48448-1G
Tris(2,2,3,3,3-pentafluoropropyl) borate
Tris(2,2,3,3,3-pentafluoropropyl) borate
Vzorec: C₉H₆BF₁₅O₃
Bod varu: 110 °C (220 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD22124053
CAS číslo: 2003-04-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR