Tisk…

Tris(1H,1H-heptafluorobutyl) borate

Dodavatel: Apollo Scientific
Varování

Synonyms: Tris(2,2,3,3,4,4,4-heptafluorobut-1-yl)boron, Tris(2,2,3,3,4,4,4-heptafluorobutoxy)borane, Tris(2,2,3,3,4,4,4-heptafluorobut-1-yl) borate

PC48447-5G PC48447-1G
APOSPC48447-5GEA 17300 CZK
APOSPC48447-5G APOSPC48447-1G
Tris(1H,1H-heptafluorobutyl) borate
Tris(1H,1H-heptafluorobutyl) borate
Vzorec: C₁₂H₆BF₂₁O₃
MW: 607,952 g/mol
Bod varu: 137 °C (200 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD22124054
CAS číslo: 755-53-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR