Tisk…

3-(Difluoromethoxy)piperidine 95%

Dodavatel: Apollo Scientific
Varování

PC48445-1G PC48445-250MG PC48445-500MG
APOSPC48445-1GEA 65130 CZK
APOSPC48445-1G APOSPC48445-250MG APOSPC48445-500MG
3-(Difluoromethoxy)piperidine 95%
3-(Difluoromethoxy)piperidine
Vzorec: C₆H₁₁F₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD24843076
CAS číslo: 1240528-91-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR