Tisk…

6-Bromo-3-chloro-2-fluorobenzoic acid 97%

Dodavatel: Apollo Scientific
Varování

PC48441-1G PC48441-250MG PC48441-5G
APOSPC48441-1GEA 6020 CZK
APOSPC48441-1G APOSPC48441-250MG APOSPC48441-5G
6-Bromo-3-chloro-2-fluorobenzoic acid 97%
6-Bromo-3-chloro-2-fluorobenzoic acid
Vzorec: C₇H₃BrClFO₂
MW: 253,453 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23713162
CAS číslo: 1428234-67-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR