Tisk…

Methyl 2-fluoro-5-(hydroxymethyl)benzoate 96%

Dodavatel: Apollo Scientific
Varování

Synonyms: [4-Fluoro-3-(methoxycarbonyl)phenyl]methanol, 4-Fluoro-3-(methoxycarbonyl)benzyl alcohol

PC48437-1G PC48437-250MG PC48437-5G
APOSPC48437-1GEA 6220 CZK
APOSPC48437-1G APOSPC48437-250MG APOSPC48437-5G
Methyl 2-fluoro-5-(hydroxymethyl)benzoate 96%
Methyl 2-fluoro-5-(hydroxymethyl)benzoate
Vzorec: C₉H₉FO₃
MW: 184,164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19381748
CAS číslo: 816449-70-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR