4'-Fluoro-3'-nitroacetanilide 98%
Dodavatel: Apollo Scientific
Synonyms:
5-Acetamido-2-fluoronitrobenzene, 5-(Acetylamino)-2-fluoronitrobenzene, N-(4-Fluoro-3-nitrophenyl)acetamide
PC48425-1G
PC48425-5G
APOSPC48425-1GEA
1370
CZK
APOSPC48425-1G
APOSPC48425-5G
4'-Fluoro-3'-nitroacetanilide 98%
4'-Fluoro-3'-nitroacetanilide
Vzorec:
C₈H₇FN₂O₃ MW: 198,151 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00585083 CAS číslo: 351-32-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...