Tisk…

3-Hydroxy-5-(trifluoromethyl)benzonitrile

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Cyano-5-(trifluoromethyl)phenol, 3-Cyano-5-hydroxybenzotrifluoride

PC48422-1G PC48422-250MG PC48422-5G
APOSPC48422-1GEA 14490 CZK
APOSPC48422-1G APOSPC48422-250MG APOSPC48422-5G
3-Hydroxy-5-(trifluoromethyl)benzonitrile
3-Hydroxy-5-(trifluoromethyl)benzonitrile
Vzorec: C₈H₄F₃NO
MW: 187,119 g/mol
Bod tání: 128…130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16999150
CAS číslo: 1243459-56-4
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR