Tisk…

3-Bromo-6-fluoro-2-nitrotoluene

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Bromo-3-fluoro-2-methylnitrobenzene, 1-Bromo-4-fluoro-3-methyl-2-nitrobenzene

PC48413-1G PC48413-25G PC48413-5G
APOSPC48413-1GEA 1490 CZK
APOSPC48413-1G APOSPC48413-25G APOSPC48413-5G
3-Bromo-6-fluoro-2-nitrotoluene
3-Bromo-6-fluoro-2-nitrotoluene
Vzorec: C₇H₅BrFNO₂
MW: 234,023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18632532
CAS číslo: 1286734-82-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR