Tisk…

2-Fluoro-5-(trifluoromethyl)styrene 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Fluoro-3-vinylbenzotrifluoride, 2-Ethenyl-1-fluoro-4-(trifluoromethyl)benzene

PC48408-1G PC48408-5G
APOSPC48408-1GEA 3310 CZK
APOSPC48408-1G APOSPC48408-5G
2-Fluoro-5-(trifluoromethyl)styrene 95%
2-Fluoro-5-(trifluoromethyl)styrene
Vzorec: C₉H₆F₄
MW: 190,138 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08461649
CAS číslo: 1286734-95-9
UN: 1993
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR