Tisk…

1-[3-Nitro-5-(trifluoromethyl)phenyl]ethanone 95%

Dodavatel: Apollo Scientific
Varování

PC48398-1G PC48398-250MG PC48398-5G
APOSPC48398-1GEA 1040 CZK
APOSPC48398-1G APOSPC48398-250MG APOSPC48398-5G
1-[3-Nitro-5-(trifluoromethyl)phenyl]ethanone 95%
1-[3-Nitro-5-(trifluoromethyl)phenyl]ethanone
Vzorec: C₉H₆F₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD11847337
CAS číslo: 39174-87-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR