2-Fluoro-5-(1,1,1,2,3,3,3-heptafluoropropan-2-yl)benzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
PC48395-1G
PC48395-250MG
APOSPC48395-1GEA
2690
CZK
APOSPC48395-1G
APOSPC48395-250MG
2-Fluoro-5-(1,1,1,2,3,3,3-heptafluoropropan-2-yl)benzaldehyde
2-Fluoro-5-(1,1,1,2,3,3,3-heptafluoropropan-2-yl)benzaldehyde
Vzorec:
C₁₀H₄F₈O Storage Temperature: Ambient |
MDL Number:
MFCD31812095 CAS číslo: 2379918-42-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...