2-Bromo-5-fluoro-3-nitrobenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC48391-1G
PC48391-250MG
PC48391-5G
APOSPC48391-1GEA
705
CZK
APOSPC48391-1G
APOSPC48391-250MG
APOSPC48391-5G
2-Bromo-5-fluoro-3-nitrobenzoic acid
2-Bromo-5-fluoro-3-nitrobenzoic acid
Vzorec:
C₇H₃BrFNO₄ Storage Temperature: Ambient |
MDL Number:
MFCD12173018 CAS číslo: 1055331-73-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...