4-Bromo-1,2-bis(trifluoromethyl)benzene 95+%
Dodavatel: Apollo Scientific
PC48379-1G
PC48379-250MG
APOSPC48379-1GEA
7840
CZK
APOSPC48379-1G
APOSPC48379-250MG
4-Bromo-1,2-bis(trifluoromethyl)benzene 95+%
4-Bromo-1,2-bis(trifluoromethyl)benzene
Vzorec:
C₈H₃BrF₆ MW: 293,004 g/mol Bod varu: 160…161 °C Bod tání: –48…–47 °C Storage Temperature: Ambient |
MDL Number:
MFCD06253758 CAS číslo: 320-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...