4-(Difluoromethyl)benzonitrile
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
α,α-Difluoro-p-tolunitrile, 4-Cyanobenzal fluoride
PC48367-1G
PC48367-250MG
APOSPC48367-1GEA
5790
CZK
APOSPC48367-1G
APOSPC48367-250MG
4-(Difluoromethyl)benzonitrile
4-(Difluoromethyl)benzonitrile
Vzorec:
C₈H₅F₂N MW: 153,129 g/mol Bod varu: 80…90 °C (20 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD23843788 CAS číslo: 55805-10-2 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...