Tisk…

4-Nitro-2-(trifluoromethyl)benzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(Bromomethyl)-4-nitro-2-(trifluoromethyl)benzene, 2-(Bromomethyl)-5-nitrobenzotrifluoride, 1-(Bromomethyl)-2-(trifluoromethyl)-4-nitrobenzene

PC48366-1G PC48366-5G
APOSPC48366-1GEA 6220 CZK
APOSPC48366-1G APOSPC48366-5G
4-Nitro-2-(trifluoromethyl)benzyl bromide 97%
4-Nitro-2-(trifluoromethyl)benzyl bromide
Vzorec: C₈H₅BrF₃NO₂
MW: 284,03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847129
CAS číslo: 694499-22-4
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR