1,2-Dichloro-1,1,2-trifluoro-4-iodobutane
Dodavatel: Apollo Scientific
PC4836-25G
PC4836-5G
APOSPC4836-25GEA
10350
CZK
APOSPC4836-25G
APOSPC4836-5G
1,2-Dichloro-1,1,2-trifluoro-4-iodobutane
1,2-Dichloro-1,1,2-trifluoro-4-iodobutane
Vzorec:
C₄H₄Cl₂F₃I MW: 306,88 g/mol Bod varu: 65 °C (25 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00042242 CAS číslo: 679-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...