Tisk…

4-Nitro-2-(trifluoromethyl)benzaldehyde

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Formyl-5-nitrobenzotrifluoride, 4-Nitro-α,α,α-trifluoro-o-tolualdehyde

PC48359-1G PC48359-5G
APOSPC48359-1GEA 622 CZK
APOSPC48359-1G APOSPC48359-5G
4-Nitro-2-(trifluoromethyl)benzaldehyde
4-Nitro-2-(trifluoromethyl)benzaldehyde
Vzorec: C₈H₄F₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD13185769
CAS číslo: 50551-17-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR