Tisk…

3,4-Dibromo-5-fluorobenzotrifluoride 95%

Dodavatel: Apollo Scientific
Varování

PC48349-1G PC48349-250MG PC48349-5G
APOSPC48349-1GEA 1660 CZK
APOSPC48349-1G APOSPC48349-250MG APOSPC48349-5G
3,4-Dibromo-5-fluorobenzotrifluoride 95%
3,4-Dibromo-5-fluorobenzotrifluoride
Vzorec: C₇H₂Br₂F₄
MW: 321.892 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28784690
CAS číslo: 1804417-25-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR