Methyl 3-bromo-2,5-difluorobenzoate 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC48338-250MG
PC48338-1G
APOSPC48338-250MGEA
12200
CZK
APOSPC48338-250MG
APOSPC48338-1G
Methyl 3-bromo-2,5-difluorobenzoate 95%
Methyl 3-bromo-2,5-difluorobenzoate
Vzorec:
C₈H₅BrF₂O₂ MW: 251,025 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24107103 CAS číslo: 1524902-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...