Tisk…

2-Fluoro-5-methoxybenzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC48335-1G PC48335-250MG PC48335-5G
APOSPC48335-1GEA 3110 CZK
APOSPC48335-1G APOSPC48335-250MG APOSPC48335-5G
2-Fluoro-5-methoxybenzyl bromide 97%
2-Fluoro-5-methoxybenzyl bromide
Vzorec: C₈H₈BrFO
Bod varu: 130…138 °C (18 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11617285
CAS číslo: 91319-42-5
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR