Tisk…

3-Amino-2-methylphenyl trifluoromethanesulphonate

Dodavatel: Apollo Scientific
Nebezpečí

PC48334-1G PC48334-250MG
APOSPC48334-1GEA 6950 CZK
APOSPC48334-1G APOSPC48334-250MG
3-Amino-2-methylphenyl trifluoromethanesulphonate
3-Amino-2-methylphenyl trifluoromethanesulphonate
Vzorec: C₈H₈F₃NO₃S
Storage Temperature: Ambient
MDL Number: MFCD26407437
CAS číslo: 1820706-98-6
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR