Tisk…

1-Bromo-7-fluoroisoquinoline

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Bromo-7-fluoro-2-azanaphthalene

PC48317-500MG PC48317-100MG PC48317-250MG
APOSPC48317-500MGEA 6250 CZK
APOSPC48317-500MG APOSPC48317-100MG APOSPC48317-250MG
1-Bromo-7-fluoroisoquinoline
1-Bromo-7-fluoroisoquinoline
Vzorec: C₉H₅BrFN
MW: 226,045 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15832812
CAS číslo: 1207448-24-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR