Tisk…

2-(Difluoromethoxy)-4-fluorobromobenzene 98%

Dodavatel: Apollo Scientific

PC48301-100G PC48301-25G PC48301-5G
APOSPC48301-100GEA 46290 CZK
APOSPC48301-100G APOSPC48301-25G APOSPC48301-5G
2-(Difluoromethoxy)-4-fluorobromobenzene 98%
2-(Difluoromethoxy)-4-fluorobromobenzene
Vzorec: C₇H₄BrF₃O
Storage Temperature: Refrigerator
MDL Number: MFCD11036070
CAS číslo: 954235-83-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR